Imprimer
Millipore® Immobilon®-NC Nitrocellulose Transfer Membranes
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HATF304F0 HATF00010
MILFHATF304F0EA 548 EUR
MILFHATF304F0 MILFHATF00010
Millipore® Immobilon®-NC Nitrocellulose Transfer Membranes
Membranes de transfert
Immobilon-NC (HATF) membrane is composed of a mixed cellulose esters matrix. It is devoid of surfactants that may interfere with cell wall integrity during cell growth
  • Can be used in western blot analysis
  • Quality level: 400
This membrane is recommended for colony and plaque lifts.