Imprimer
TriLink® CleanCap® M6 Cas9 mRNA (N1MePsU)
Marques : TriLink BioTechnologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
L-8106-100 L-8106-1000 L-8106-5
SOLUL-8106-100EA 388 EUR
SOLUL-8106-100 SOLUL-8106-1000 SOLUL-8106-5
TriLink® CleanCap® M6 Cas9 mRNA (N1MePsU)
Protéines expression Réactifs pour l'expression protéique
Cas9 mRNA expresses a version of the Streptococcus pyogenes SF370 Cas9 protein (CRISPR Associated Protein 9). Cas9 functions as part of the CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) genome editing system. In the CRISPR system, an RNA guide sequence targets the site of interest and the Cas9 protein is employed to perform double-stranded DNA cleavage.
  • Capped with CleanCap® M6 cap analog, modified with N1-methylpseudouridines to enhance protein expression
  • Cap 1 structure with >95% capping efficiency for superior in vivo activity compared to Cap-0 mRNAs
  • Manufactured with CleanCap M6–specific IVT that lowers dsRNA and improves performance
  • Ready to use in downstream mRNA applications and formulations
  • Grade: RUO
This mRNA is capped using CleanCap® Reagent M6, TriLink’s patented co-transcriptional capping technology, resulting in the naturally occurring Cap-1 structure with >95% capping efficiency. It is polyadenylated, modified with N1-methylpseudouridine, and optimised for mammalian systems. It mimics a fully processed mature mRNA.
CleanCap Reagent M6, otherwise known as CleanCap m6AG 3′ OMe, produces a base-modified Cap-1 mRNA, which shows superior in vivo activity compared to Cap-0 mRNA produced by legacy capping methods such as mCap or anti-reverse cap analog (ARCA). CleanCap M6 may further increase protein expression relative to previous generations of cap analogs, such as CleanCap AG or CleanCap AG (3′ OMe), or mRNAs produced by enzymatic capping strategies.
N1-methylpseudouridine is a modified uridine that can reduce immunogenic response and enhance translational efficiency of mRNAs. These properties can result in safer mRNA and increased protein expression.
Attention: Item will be delivered at or below –40 °C and should be stored at or below –40 °C.