Imprimer
TriLink® CleanCap® Renilla Luc mRNA (5 moU)
Marques : TriLink BioTechnologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
L-7204-1000 L-7204-100 L-7204-5
SOLUL-7204-1000EA 1760 EUR
SOLUL-7204-1000 SOLUL-7204-100 SOLUL-7204-5
TriLink® CleanCap® Renilla Luc mRNA (5 moU)
Protéines expression Réactifs pour l'expression protéique
Renilla luciferase (Renilla Luc) is a bioluminescent reporter protein derived from Renilla reniformis (sea pansy). In the presence of the cofactor, coelenterazine, Renilla luciferase produces light. Renilla Luc mRNA can be used as an internal control in combination with FLuc mRNA to test different conditions, such as dosage or chemical modifications, and their effect on translation.
  • Capped with CleanCap® AG cap analog, modified with 5-methoxyuridines
  • Cap 1 structure with >95% capping efficiency for superior in vivo activity compared to Cap-0 mRNAs
  • Manufactured with TriLink’s proprietary CleanScript® method to lower dsRNA for improved performance
  • Ready to use in downstream mRNA applications and formulations
  • Grade: RUO
This mRNA is capped using CleanCap® Reagent AG, TriLink's co-transciptional capping technology, which results in the naturally occurring Cap-1 structure with >95% capping efficiency. It is manufactured using TriLink’s CleanScript® method, polyadenylated, modified with 5-methoxyuridine, and optimised for mammalian systems. It mimics a fully processed mature mRNA.
CleanCap Reagent AG produces a base-modified Cap-1 mRNA, which shows superior in vivo activity compared to Cap-0 mRNA produced by legacy capping methods such as mCap or anti-reverse cap analog (ARCA).
5-methoxyuridine (5moU) is a modified uridine that can reduce immunogenic response and enhance translational efficiency of mRNAs. These properties can result in safer mRNA and increased protein expression.
CleanScript method is TriLink’s proprietary in vitro transcription process that is optimised to minimise dsRNA formation and increase mRNA activity in vivo.
Attention: Item will be delivered at or below −40 °C and should be stored at or below −40 °C.