Imprimer
Gel de silice HPTLC 60 Å, greffé, plaques de verre
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.05914.0001 1.13124.0001 1.13724.0001 1.13725.0001 1.13726.0001 1.14296.0001 1.16225.0001
1.05914.0001EA 1200 EUR
1.05914.0001 1.13124.0001 1.13724.0001 1.13725.0001 1.13726.0001 1.14296.0001 1.16225.0001
Gel de silice HPTLC 60 Å, greffé, plaques de verre
Plaques CCM
Our NH₂, CN- and DIOL-modified silica sorbents are less polar then conventional silica phases, making them ideal for separating hydrophilic or charged substances. Our moderately polar cyano- and diol-modified silica plates can be used for both normal phase and reversed phase systems. An alternative to PEI cellulose, amino-modified NH₂ plates provide weak basic ion exchange characteristics with special selectivity for charged compounds.
HPTLC concentrating zone plates allow easy application of large volumes of diluted samples. They include a purification and concentration step.