Imprimer
HPTLC plates, nano silica gel 60, unmodified layers, Nano-SIL HD
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
811213 811223 811211 811212 811221 811222
MANA811213EA 679 EUR
MANA811213 MANA811223 MANA811211 MANA811212 MANA811221 MANA811222
HPTLC plates, nano silica gel 60, unmodified layers, Nano-SIL HD
Plaques CCM
Unmodified HPTLC silica layers with "High Definition" properties for HPTLC.
  • Nano silica gel (particle size 2 to 10 μm)
  • Optimised binder system
  • High luminosity and brilliant staining properties
Besides a good wettability for precise colorisation results the Nano-SIL HD glass plates allows sharper separations, shorter developing times and shorter migration distances compared to standard silica layers. High suitability for trace analyses resulting from a UV indicator with increased brilliance and a low-noise background of the layer.
Nano silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 2 – 10 μm.