Imprimer
TLC plates, silica gel 60, unmodified layers, SIL HD
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
809213 809223 809210 809212 809217 809220 809222 809227
MANA809213EA 286 EUR
MANA809213 MANA809223 MANA809210 MANA809212 MANA809217 MANA809220 MANA809222 MANA809227
TLC plates, silica gel 60, unmodified layers, SIL HD
Plaques CCM
Unmodified standard silica layers with "High Definition" properties for TLC.
  • High-quality silica gel
  • Optimised binder system
  • High luminosity and brilliant staining properties
SIL HD glass plates are characterised by good wettability for precise colorisation results, even with 100% aqueous detection reagents. High suitability for trace analyses resulting from a UV indicator with increased brilliance and a low-noise background of the layer.
Silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 5 – 17 μm.