Imprimer
Freund's incomplete adjuvant, IFAVax
Marques : OZ BIOSCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
IFA0010 IFA0050 IFA0100
OZBIIFA0010EA 47.59 EUR
OZBIIFA0010 OZBIIFA0050 OZBIIFA0100
Freund's incomplete adjuvant, IFAVax
Protéines expression Réactifs pour l'expression protéique
Incomplete Freund’s Adjuvant (IFA) is a water-in-oil emulsion that is similar to the complete Freund’s Adjuvant (CFA) without addition of heat-killed mycobacteria (Mycobacterum butyricum).
  • Suitable for the stimulation of Th2 response and antibody production
It consists of a mixture of mineral oil and emulsifier in a ratio of 85% v/v oil and 15% v/v emulsifier. It is not a pre-formed emulsion and thus it must be mixed with an equal volume of aqueous solution of antigen and subsequently emulsified prior to use. The emulsifier is mannide monooleate, an ester consisting of mannitol as the hydrophilic residue and oleic acid (unsaturated C18 fatty acid) as the hydrophobic part.
Store at +4 °C
Informations de livraison: Shipped at room temperature.
Attention: Intended for research use only.