Imprimer
HPTLC plates silica gel 60, modified CN, DIOL, NH₂, glass backed
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.12668.0001 1.13192.0001 1.15647.0001 1.16464.0001 1.05636.0001
1.12668.0001EA 610 EUR
1.12668.0001 1.13192.0001 1.15647.0001 1.16464.0001 1.05636.0001
HPTLC plates silica gel 60, modified CN, DIOL, NH₂, glass backed
TLC Plates
CN- and DIOL-modified silica plates are moderately polar that are suitable for both normal and reversed phase systems. The amino modified NH₂ plate provides weak basic ion exchange characteristics with special selectivity for charged compounds offering an alternative to PEI cellulose.