Imprimer
Membranes de transfert, nitrocellulose, Amersham™ Protran®
Fournisseur: Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
10600033 10600044 10600041 10600042 10600032 10600043 10600046 10600045 10600012 10600000 10600001 10600002 10600005 10600006 10600007 10600011 10600010 10600093 10600094 10600114 10600115 10600116 10600062 10600073 10600125 10600124
10600033.EA 448 EUR
10600033. 10600044. 10600041. 10600042. 10600032. 10600043. 10600046. 10600045. 10600012. 10600000. 10600001. 10600002. 10600005. 10600006. 10600007. 10600011. 10600010. 10600093. 10600094. 10600114. 10600115. 10600116. 10600062. 10600073. 10600125. GEHE10600124
Membranes de transfert, nitrocellulose, Amersham™ Protran®
Membranes de transfert
Amersham™ Protran® membranes are manufactured from 100% pure nitrocellulose to ensure the highest binding capacity possible. They are compatible with a variety of detection methods, including isotopic, chemiluminescent, colorimetric and fluorescent. No methanol pre-wetting step is required, making this the membrane of choice for proteins which prefer aqueous environments. Prior to transfer the membrane is simply wetted with water, and then placed in the transfer buffer. No other pretreatment steps are necessary.
  • Proprietary nitrocellulose formula gives a long shelf life of bound proteins
  • High binding capacity
  • Low background and excellent signal-to-noise ratios
  • Compatible with a variety of labelling and detection systems
  • Available in popular pre-cut sizes
Amersham™ Protran® offers excellent sensitivity, resolution, and low background, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western Blotting Systems.
Available in a choice of pore sizes: Protran® 0.1 NC (pore size 0,1 µm), Protran® 0.2 NC (pore size 0,2 µm), or Protran® 0.45 NC (pore size 0,45 µm).
The versatile Protran® 0.1 NC membrane (binding capacity 231 to 248 µg IgG/cm²) provides excellent binding affinity for small proteins and peptides, as well as nucleic acids.
Protran® 0.2 NC (binding capacity 150 to 176 µg IgG/cm²) binds small proteins (
Protran® 0.45 NC (binding capacity 162 to 180 µg IgG/cm²) is suitable for proteins of a wide range of molecular weights.
For convenience, Western blotting sandwiches (10 sandwiches/package) are also available. Each sandwich consists of pre-cut Amersham™ Protran® NC membranes pre-assembled with 2×3 mm Chr filter papers.