Imprimer
TLC plates, cellulose MN 300 PEI, polyester sheets, POLYGRAM®
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
801053 801063
552-0136EA 296 EUR
552-0136 552-0139
TLC plates, cellulose MN 300 PEI, polyester sheets, POLYGRAM®
Plaques CCM
Fibrous cellulose impregnated with polyethyleneimine available as POLYGRAM® polyester sheets with or without fluorescent indicator. They are used in analysis of nucleic acids and of mutagenic substances with the ³²P post labelling procedure.
  • Ion exchange layer for TLC
Base material: Celluolse MN 300: Fibre length (95%) 2 to 20 μmg; average degree of polymerisation 400 to 500; specific surface acc. to Blaine 15000 cm²/g.
Impregnation: Polyethyleneimine (PEI).