Imprimer
Plaques CCMHP, gel de silice 60, couches non modifiées, revêtement en verre, Nano-ADAMANT
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
821110 821120 821140 821150
552-0344EA 276 EUR
552-0344 552-0345 552-0347 552-0348
Plaques CCMHP, gel de silice 60, couches non modifiées, revêtement en verre, Nano-ADAMANT
Plaques CCM
Unmodified HPTLC silica layers available as glass plates with or without fluorescent indicator. The narrow fractionation of the silica particles features theoretical plate heights, which are one order of magnitude smaller than on standard silica layers. Thus, Nano-ADAMANT has the advantage of sharper separations, shorter developing times, and increased detection sensitivity with equal selectivity.
  • Good hardness and abrasion resistance due to an optimized binder system
  • Increased separation efficiency due to an optimised particle size distribution
  • High suitability for trace analyses resulting from a UV indicator with increased brilliance and a low-noise background of the layer
Nano silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 2 to 10 µm.