Imprimer
Plaques CCMHP, gel de silice 60, couches non modifiées, revêtement en verre, Nano-DURASIL
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
812010 812011 812013 812014
552-0267EA 276 EUR
552-0267 552-0268 552-0270 552-0271
Plaques CCMHP, gel de silice 60, couches non modifiées, revêtement en verre, Nano-DURASIL
Plaques CCM
These unmodified HPTLC silica layers are available as glass plates with or without fluorescent indicator. The narrow fractionation of the silica particles features sharper separations, shorter developing times, and an increased detection sensitivity compared to DURASIL plates.
  • Hard and water-resistant layers due to a special binder system
  • Different selectivity compared to Nano-SIL plates
Nano silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 2 to 10 μm.