Imprimer
Plaques CCM, gel de silice 60, couches non modifiées, SIL G
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
805012 805013 805021 805022 805023 809011 809012 809013 809022 809023 809033 809043 809121 809122 809123 818033 818132 818133 818131 818030.20 818032 818130.20 818160 818161 818162 818163 805014 805017 805024 805032 809017 809020 809021 809027 809051 809053 809061 809063 809073 809083 810063 809017.200 809027.200 809028.100 805901 805902 818129
552-0157EA 117 EUR
552-0157 552-0158 552-1291 552-0161 552-0162 552-0190 552-0191 552-0192 552-0197 552-0198 552-0202 552-0203 552-0210 552-0211 552-0212 552-0295 552-0298 552-0299 552-0297 552-0293 552-0294 552-0296 552-0314 552-0315 552-0316 552-0317 552-0159 552-0160 552-0163 552-0164 552-0193 552-0195 552-0196 552-0199 552-0204 552-0205 552-0206 552-0207 552-0208 552-0209 552-0220 552-0194 552-0200 552-0201 MANA805901 MANA805902 MANA818129
Plaques CCM, gel de silice 60, couches non modifiées, SIL G
Plaques CCM
Unmodified standard silica layers for TLC.
  • High-quality silica gel
  • Indicator F254 or F366
  • Binders are stable in almost all organic solvents
These highly robust plates and flexible sheets with gypsum and polymeric binder systems are well-suitable for every standard application.
SIL G glass plates with a layer thickness from 500 to 2000 µm are used for preparative TLC.
SIL G-25 HR plates are coated with high purity silica 60, bonded with gypsum and a very small quantity of a polymeric organic binder. They are recommended for aflatoxin separations.
SIL G-25 Tenside plates are impregnated with ammonium sulphate and are applied for separations of surfactants.
Silica 60: Specific surface (BET) ~ 500 m2/g; specific pore volume 0,75 ml/g; particle size 5 to 17 μm.