Imprimer
Squalene emulsion, SqualVax
Marques : OZ BIOSCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
SQ0010 SQ0050 SQ0100
OZBISQ0010EA 176.79 EUR
OZBISQ0010 OZBISQ0050 OZBISQ0100
Squalene emulsion, SqualVax
Protéines expression Réactifs pour l'expression protéique
SqualVax is an oil-in-water emulsion made of squalene droplets in a continuous aqueous phase. It is an important advantage over alternative oils that have been used in emulsion adjuvants, like Freund’s adjuvant that contains mineral oil (paraffin oil) and has long term persistence in organisms.
Squalene emulsion induces local stimulation and recruitment of DCs and granulocytes, differentiation of monocytes into DCs and increased uptake of antigen by DCs. The emulsion acts more specifically on macrophages present at the site of injection. This formulation enhances differentiation of monocytes towards a mature phenotype, thereby promoting migration of antigen-loaded cells to the draining lymph node.
Informations de livraison: Shipped at room temperature.
Cdt: Store at +4 °C.