Imprimer
Transfer membranes, PVDF, Owl™, Immobilon-P™
Fournisseur: THERMO OWL SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
TM151-1 TM151-4 TM151-5
OWLATM151-1EA 753 EUR
OWLATM151-1 OWLATM151-4 OWLATM151-5
Transfer membranes, PVDF, Owl™, Immobilon-P™
Membranes de transfert
Hydrophobic polyvinylidene fluoride transfer membrane has a high signal-to-noise ratio, which allows for the use of mild blocking procedures
Membrane has high protein retention, is compatible with all standard visualization methods, and can be used for protein sequencing. Available in sheet or roll form.
Thermo Scientific offers Immobilon™-P, PVDF transfer membrane manufactured by Millipore™