Imprimer
Affinity protein expression and purification systems
Marques : AGILENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
214303
AGLS214303EA 450 EUR
AGLS214303
Affinity protein expression and purification systems
Protéines expression Systèmes d'expression des protéines
The affinity protein expression and purification system provides a simple, gentle and effective method for high-level protein expression and purification.
  • pCAL vectors derived from T7 RNA polymerase-based pET vectors
  • Novel purification tag combines small size, gentle elution and high-affinity binding
  • Easy tag removal resulting in native protein
Informations de livraison: The complete system includes pCAL expression vectors containing a calmodulin-binding peptide tag and calmodulin affinity resin (available separately).
Attention: For research use only. Not for use in diagnostic procedures.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable