Imprimer
Gas Washing Bottles, Head with VitraPOR® Filter Disc, Standard Joint NS 34/35
Fournisseur: ROBU GLASFILTER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
34101 34100 34102
201-1461EA 171 EUR
201-1461 ROBU34100 ROBU34102
Gas Washing Bottles, Head with VitraPOR® Filter Disc, Standard Joint NS 34/35
Flacons Flacons-laveurs
ROBU® gas washing bottle head with integrated VitraPOR® filter disc and standard ground joint ensures efficient gas-liquid contact and uniform dispersion in absorption processes.
  • High-quality borosilicate glass construction
  • Includes a VitraPOR® filter disc for efficient filtration of gases
  • Standard joint for easy connection to other laboratory equipment NS 34/35
  • Resistant to chemicals and high temperatures
  • Ideal for gas washing and purification
  • Ensures reliable filtration and separation of impurities
  • Compatible with standard laboratory setups
The ROBU gas washing bottle head with VitraPOR filter disc and standard joint is designed for efficient gas scrubbing in laboratory setups. The integrated filter disc ensures fine dispersion of gases into the washing liquid, maximising surface contact and absorption efficiency.
Made from high-quality borosilicate glass, it offers excellent chemical resistance and durability. The standardised joint ensures a secure, leak-free fit with compatible gas washing bottles. Ideal for removing impurities, neutralising aggressive gases, or humidifying gas streams, the ROBU head component guarantees reliable performance in chemical processing and analytical applications.
Certifications:
In compliance with ISO 4796-80 and ASTM E128-99 standards.