Imprimer
WEDO® Super Step Kick Stools
Fournisseur: WEDO WERNER DORSCH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
212300 212301 212302 212303 212312 212337
WEDO212300EA 118 EUR
WEDO212300 WEDO212301 WEDO212302 WEDO212303 WEDO212312 WEDO212337
WEDO® Super Step Kick Stools
Escabeaux et marchepieds
The WEDO® Super Step castor stool is made of unbreakable, lightfast polypropylene with treads made of TPE.
  • Unbreakable
  • Rollable
  • Non-slip treads
The slip resistance of the treads is improved by the joint processing of the two components so that slipping is practically impossible.
The treads cannot be removed. As both plastics come from the same family, the Super Step body can still be 100% recycled and is assembled by simply plugging the upper and lower parts together. No tools are required.
The three replaceable castors mounted on steel springs are pre-assembled. Under load, the Super Step® stands firmly on its integrated rubber support ring.
Max. load capacity (tested and approved): 150 kg.
Certifications:
TÜV and GS tested in accordance with EN 14183-F.
Informations de commande:
Ten years guarantee on the plastic body.