Imprimer
VitraPOR® Filter Candle-Blanks, Conical Shape
Fournisseur: ROBU GLASFILTER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
18201 18202 18203 18204 18212 18213 18214 18210 18200 18211
511-0201EA 103 EUR
511-0201 511-0202 511-0203 511-0204 ROBU18212 ROBU18213 ROBU18214 ROBU18210 ROBU18200 ROBU18211
VitraPOR® Filter Candle-Blanks, Conical Shape
Filtres Tubes filtrants
VitraPOR® conical filter candle-blanks are ideal for custom filter assemblies. The conical shape ensures secure fitting, while the porous glass delivers precise and reliable filtration performance.
  • VitraPOR® porous glass filter
  • Conical shape for tight fit
  • Customisable for various systems
  • High chemical and thermal resistance
  • Suitable for gas and liquid filtration
  • Reusable and autoclavable
  • Uniform pore structure
ROBU VitraPOR filter candle-blanks in conical shape are designed for flexible integration into filtration setups where a press-fit or sealed connection is required. Made of borosilicate glass 3.3 with a homogeneous structure, they ensure reliable filtration of gases and liquids with excellent chemical and thermal durability. These blanks serve as a robust base for custom filter solutions and can be regenerated by thermal treatment or cleaning. Ideal for laboratory, pilot, or production applications requiring compact and efficient filtration elements.
Certifications:
In compliance with ISO 4796-80 and ASTM E128-99 standards.