Imprimer
ScienceLine plus pH Electrodes
Fournisseur: SI Analytics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
285101310 285101345
SCOI285101310EA 705 EUR
SCOI285101310 SCOI285101345
ScienceLine plus pH Electrodes
Électrodes Électrodes pH
ScienceLine Plus pH electrodes with Silamid®-reference system with silver ion barrier and double electrolyte.
  • Faster and more stable results and a longer electrode lifespan
  • Higher accuracy than traditional analog sensors
  • Effortless capture, storage of your sensors latest calibration data
  • Highest possible operator comfort and measuring precision
pH electrodes that use chlorinated silver wire to form a silver/silver chlorine reference system (diaphragm) that degrades over time when in contact with certain sample types, all ScienceLine Plus series electrodes employ a Silamid® double reference with silver-ion trap that facilitates universal use, even in protein or sulphide containing samples.
The diaphragm itself is a maintenance free encapsulated gel system, and the silver coating of the inner tube has significantly larger (5x) silver surface compared to the silver wire system making for a faster, more stable and longer. The outer bridge electrolyte consists of the proven 3 mol/l KCl.
IDS digital electrodes facilitate connection to IDS compatible meters or digital gateway such as the IDS gate. A glass model for universal application (DIN or IDS connector). Sensor function of pH and temperature.
SCP A 170 MF ID: Electrode with sphere membrane with large surface area.
SCP MIC AMF IDS: Micro electrode for small probes.
Certifications:
GLP or FDA regulation 21 CFR Part 11