Imprimer
VWR® Glass Vials with Plastic Snap-On Lid
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
216-1758EA 47.5 EUR
216-1758 216-1759 216-1760 216-1770 216-1776 216-1777 216-1778 216-1782 216-1843 216-1844 216-1845 216-1846 216-1847 216-1848 216-1849 216-1851
VWR® Glass Vials with Plastic Snap-On Lid
Fioles
Roll rim jars are ideal for storing, transporting, and protecting samples, chemicals, or liquids.
  • Ideal for safely storing and transporting samples, chemicals, or liquids
  • Clear glass offers the advantage of clearly visible contents
  • Amber glass offers additional protection from light; particularly important for light-sensitive substances
  • Both glass variants are robust and reusable
The snap-on lids ensure a secure closure and prevents contents from leaking out. The different types of glass, clear or amber, offer protection against light or chemical reaction, depending on the application.