Imprimer
epMotion® 96 Flex Dispensing Heads
Fournisseur: EPPENDORF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5069000.527 5069000.535
EPPE5069000.527EA 7750 EUR
EPPE5069000.527 EPPE5069000.535
epMotion® 96 Flex Dispensing Heads
Automated Liquid Handling Systems
The epMotion® 96 Flex’s exchangeable dispensing head design accommodates a very large volume range with only two dispensing heads and can handle less than 96 tips.
  • Covering 0.5 µL to 1000 µL with only two dispensing heads
  • Use less than 96 tips for full flexibilty
  • Easy exchange of dispensing heads, no tools needed
  • Including hardshell case for easy transport and storage
ISO 23783-2 calibrated.