Imprimer
DYMO® LabelWriter™ 5 Cryogenic Labels
Fournisseur: GA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
LWCS-501-WH LWCS-502-WH LWCS-504-WH LWCS-506-WH LWCS-507-WH LWCS-510-WH LWCS-503-WH LWCS-505-WH
GAINLWCS-501-WHEA 192 EUR
GAINLWCS-501-WH GAINLWCS-502-WH GAINLWCS-504-WH GAINLWCS-506-WH GAINLWCS-507-WH GAINLWCS-510-WH GAINLWCS-503-WH GAINLWCS-505-WH
DYMO® LabelWriter™ 5 Cryogenic Labels
Étiquettes
Labels for frozen surfaces are now available for printing with DYMO® LabelWriter 5 series printers.
  • Temperature range: −196 to +70 °C (−321 to +158 °F)
  • Min. application temperature: −80 °C (−112 °F)
  • Permanent adhesive
  • Core size: 25.4 mm (1")
  • Quick labeling of frozen vials and tubes (no thawing required)
  • Ideal for labeling ambient temperature as well as frozen surfaces
  • Withstands exposure to solvents, water baths, and condensation
  • Compatible with the 550, 550 Turbo, and 5XL DYMO® LabelWriters
  • Convenient to print, no need for ribbons or other ink sources
These durable, solvent-resistant cryogenic labels are designed for direct application to frozen surfaces as low as −80 °C (−112 °F), including on vials and tubes stored in ultra-low temperature freezers. These labels can also be applied to room temperature vials prior to storage in ultra-low temperature freezers, ensuring secure adhesion and durability throughout the entire freezing process. They offer a reliable way to label frozen containers without needing to thaw samples first, ensuring the integrity of valuable temperature-sensitive specimens.
The labels feature a permanent adhesive that remains secure under extreme conditions, including prolonged liquid nitrogen storage (−196 °C/−321 °F). Once applied, the labels ensure long-term preservation and stability, ideal for research institutes, clinical laboratories, biobanks, and biotechnology companies.
Additionally, these DYMO® compatible labels are resistant to a variety of lab chemicals, including detergents, alcohols (100% ethanol immersion: up to 15 minutes), acetone, xylene (surface-level contact), and disinfecting solutions. They are perfect for use in a variety of harsh lab environments.
SnapPEEL™ Labels:
The SnapPEEL™ technology included for LWCS-506 and LWCS-507 are specially designed for cryogenic applications with permanent adhesive that holds up in extreme conditions, including ultra-low temperature freezers and liquid nitrogen storage (−196 °C/−321 °F). They resist a range of chemicals such as detergents, ethanol, acetone, and xylene, ensuring reliable performance in the harshest lab environments. The included CryoSTUCK® with SnapPEEL™ technology labels, with their unique shape, consisting of an adjoined circle-and-rectangle, allows the cap and side of vials to be efficiently labeled seamlessly and effortlessly. This significantly increases labeling efficiency and reduces excessive manipulation and unnecessary contact with the adhesive.
Informations de commande:
To ensure proper adhesion, please remove excess frost prior to affixing the label. Not compatible with DYMO® LabelWriter™ 4 series models 450, 450Turbo, and 4XL.