Imprimer
DYMO® LabelWriter™ 550 Direct Thermal Printer
Fournisseur: GA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DYMO-LW550EMEA
GAINDYMO-LW550EMEAEA 304 EUR
GAINDYMO-LW550EMEA
DYMO® LabelWriter™ 550 Direct Thermal Printer
Imprimantes Label Printers
The DYMO® LabelWriter™ 550 is a direct thermal printer that does not require costly inks or toner to print high-quality text and barcodes, while its compact design allows it to fit seamlessly into any workspace.
  • Patented automatic label recognition technology ensures proper alignment of DYMO® branded labels for smooth, jam-free printing
  • Reduces misprints and print defects, saving material and cutting down on label waste
  • Direct thermal printing technology produces high-quality, 300 dpi barcodes, images, and text without the need for additional inks or toner
  • Easy-to-use DYMO Connect software (included) is compatible with either Windows 8.1 (or later) and Mac 10.14 (or later)
Patented automatic label recognition technology will automatically identify label size and type, providing quick generation of the template once labels are loaded. Users never have to waste time selecting the right template. In addition, the automatic label counter indicates how many labels remain, eliminating the risk of running out unexpectedly.
Free DYMO Connect software is included with every printer, allowing the creation of 1D and 2D barcodes along with crisp, clear text and graphics. The 550 is up to 20% faster than its predecessor, the LabelWriter™ 450, and is fully compatible with the LWCS-class of cryogenic labels.
Informations de commande:
Order includes LabelWriter™ 550 label printer, starter roll of address labels, power adapter, power cable, USB cable, and quick start guide.