Imprimer
DYMO® LabelWriter™ 550 Printing Kit
Fournisseur: GA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PKTLWCS506EMEA PKTLWCS505EMEA
GAINPKTLWCS506EMEAEA 467 EUR
GAINPKTLWCS506EMEA GAINPKTLWCS505EMEA
DYMO® LabelWriter™ 550 Printing Kit
Imprimantes Label Printers
The DYMO® LabelWriter™ 550 Printing kit provides an affordable, complete printing solution for labs on a budget. Supplied with the DYMO LabelWriter™ 550 direct thermal printer, free DYMO Connect label design/barcoding software, and two rolls of LWCS-class labels.
  • Small, lightweight direct thermal printer, eliminating the need for ink or toner
  • Direct thermal printing technology adjusts to loaded DYMO® branded labels, producing crisp barcodes, images, and text
  • Easy-to-use DYMO Connect software (included) is compatible with either Windows 8.1 (or later) and Mac 10.14 (or later)
  • LWCS class labels are suitable for quick labeling of frozen vials and tubes (no thawing required)
This kit provides users with everything needed to start printing labels for the lab.
The DYMO® LabelWriter™ 550 is a direct thermal printer that eliminates the need for costly ink or toner, while its compact design seamlessly fits into any workspace. Automatic label recognition technology quickly generates the template once labels are loaded, eliminating wasted time selecting the right template. The automatic label counter indicates how many labels remain, so users will never run out unexpectedly.
The included CryoSTUCK® labels are specially designed for cryogenic applications, allowing direct application to frozen surfaces as low as −80 °C (−112 °F) without needing to thaw samples. These durable, solvent-resistant labels feature a permanent adhesive that holds up in extreme conditions, including ultra-low temperature freezers and liquid nitrogen storage (−196 °C/−321 °F). They resist a range of chemicals such as detergents, ethanol, acetone, and xylene, ensuring reliable performance in the harshest lab environments.
DYMO® Connect software allows the creation of 1D and 2D barcodes along with crisp, clear text and graphics. Compatible with both Windows and Mac OS, this software can print in bulk from Microsoft Excel and other databases, with integrated spellcheck to reduce spelling errors.
Informations de commande:
The Printing kit includes LabelWriter™ 550 label printer, starter roll of address labels, two rolls of LWCS-505-WH or LWCS-506-WH, power adapter, power cable, USB cable, and quick start guide.