Imprimer
VWR® Autoclavable Biohazard Bags, Polypropylene, Red
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
129-4029EA 267 EUR
129-4029 129-4037 129-4044
VWR® Autoclavable Biohazard Bags, Polypropylene, Red
Sachets Sacs à déchets
Sturdy, temperature resistant bags, ideal for disposing of non hazardous waste or hazardous waste that must first be autoclaved. 
  • Autoclavable up to 135 °C maximum
  • 200 mm twist ties included allows bag to expand and vent steam during cooling
  • Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709
  • Available with differnent thickness  37,5, 50, 75 µm
  • Available with printed bag, which printed with the universal biohazard symbol
  • Available with or without temperature indicator, text 'Contents Have Been Autoclaved' turns dark to indicates successful autoclaving
  • Highly resistant to tears and punctures
Can also be used for autoclaving other products. Supplied complete with wire closures that allow expansion of the bag and ventilation when cooling.
Certifications:
ISO 9001:2015, Sedex certified.