Imprimer
VWR® Autoclavable Biohazard Bags, Polypropylene, Yellow
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
129-4035EA 144 EUR
129-4035 129-4036
VWR® Autoclavable Biohazard Bags, Polypropylene, Yellow
Sachets Sacs à déchets
Sturdy, temperature resistant bags, ideal for disposing of non hazardous waste or hazardous waste that must first be autoclaved.
  • Autoclavable up to 135 °C (275 °F) maximum
  • 20 cm twist ties included allows bag to expand and vent steam during cooling
  • Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709
  • Bag is printed with the universal biohazard symbol
  • Bag has temperature indicator, text 'Contents Have Been Autoclaved' turns dark to indicates successful autoclaving
  • Highly resistant to tears and punctures
Can also be used for autoclaving other products. Supplied complete with wire closures that allow expansion of the bag and ventilation when cooling.
Certifications:
Certifications: ISO 9001:2015, Sedex.