Imprimer
Laboratory Chest Freezers, XLT
Fournisseur: Tenak A/S
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N112001 N112002 N122001
TENAN112001EA 3490 EUR
TENAN112001 TENAN112002 TENAN122001
Laboratory Chest Freezers, XLT
Congélateurs Congélateurs de laboratoire/universels
Laboratory chest freezers with a temperature range −40 to −60 °C. The undercounter model XLT U100 has a temperature range of −25 to −65 °C.
  • Pull-down time less than 100 min for personal freezer XLT C75
  • Low cost of ownership
  • Excellent stability and reliability
  • Long-term durability
  • Castors on XLT C75 and XLT C150 models
  • Undercounter Freezer XLT U100 is stackable
  • XLT C150 is an ideal personal freezer, features an integrated handle lock
  • XLT U100 has 2 port holes and whisper quiet noise level
The XLT C75 and XLT C150 chest freezer models have low power consumption but offer excellent stability, reliability, and long-term durability. There is no maintenance required, with exception of defrosting and vacuum cleaning the compressor compartment. Also, no regular service is required and in case of a failure, repair can be performed on-site by any trained freezer service engineer. Installation is quite easy as the freezer has compact outer dimensions and will fit through all standard doors. This combined with low weight and castors makes it easy to move around.
The XLT U100 Undercounter freezer XLT U100 offers user-friendliness and small dimensions making it the perfect choice for use inside the laboratory and for personal and flexible use. With a wide temperature range from −25 to +65 °C this freezer can be used for various different applications.  The XLT freezer range is especially popular among restaurants and seafood distributors but is also used in applications such as material storage and material testing, research and development within industries like automotive, telecom, aerospace, defence and more. The marine industry uses the XLT freezers for storage, shrinking and testing of various materials. Low cost of ownership due to very limited maintenance, on-site service and low power consumption. With a small footprint, this freezer is second-to-none in saving space. Installation is quite easy as the freezer has a compact outer dimensions and will fit through all standard doors or in an elevator.
Declaration of conformity: ATEX (2014/34/EU)
Certifications:
ISO 9001, ISO 14001, ISO 45001.