Imprimer
Ultra Low Temperature Freezers
Fournisseur: Tenak A/S
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N123000 N123001
TENAN123000EA 8890 EUR
TENAN123000 TENAN123001
Ultra Low Temperature Freezers
Congélateurs Congélateurs ultra-basses températures
Ultra low laboratory freezers with a temperature range from −60 to −86 °C.
  • Pull-down time less than 85 minutes (for ULT U35) and at start up is less than 173 minutes for ULT U100
  • Benchtop model has variable speed compressor
  • Noise level is below 46 dB (for ULT U35) and below 57 dB (for ULT U100)
  • High cooling stability
  • Power consumption of 3,2 kW/24 h (for ULT U35) and 6,7 kW/24 h (for ULT U100)
  • Undercounter ULT U100 units are stackable
The ULT U35 is a small tabletop laboratory freezer for cold storage of biological samples, cell preparation, vaccines and other applications needed to be stored at a temperature range from −25 to −86 °C. The ULT U35 tabletop freezer is ideal as a personal freezer. Especially with a footprint that doesn’t take up unnecessary space in a laboratory where floor space can be limited. The ULT U35 is designed to work silently in the laboratory. It is for everyday use, and it is ideal for users who frequently need to access samples and requires a quick temperature recovery. This tabletop lab freezer puts your samples right next to your workstation. These working conditions reduces the time spent on retrieving and storing samples from an alternative and distant location in the laboratory. The handle on the ULT U35 is designed to be ergonomic, and you will have no issue opening and closing the hatch multiple times every day.
The ULT U100 undercounter freezer is the perfect personal easy-access freezer, for daily use in laboratories, hospitals or any scientific research place that requires storage of sensitive samples, such as bacteria, viruses, tissue samples or cell preparations. This upright freezer needs very little maintenance or on-site service, and it has low power consumption. With a small footprint, this freezer is second-to-none in saving space. Installation is quite easy as the freezer has compact outer dimensions and will fit through all standard doors or in an elevator.
Declaration of conformity: ATEX (2014/34/EU)
Certifications:
ISO 9001, ISO 14001, ISO 45001.