Imprimer
b.safe Exhaust Filter Adapter Justrite
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
M815-01 M817-01
BOHLM815-01EA 50.2 EUR
BOHLM815-01 BOHLM817-01
b.safe Exhaust Filter Adapter Justrite
Raccords et connecteurs
Adaptor for the use of b.safe Exhaust filters on Justrite® Disposal safety cans in order to stop solvent vapours from the escaping in the air during filling with HPLC waste.
  • Made of grey PP or black PE
  • With GL 14 thread
Adaptor for assembly on the lateral couplings of Justrite® Disposal Safety Cans, with connector to screw in a b.safe Exhaust filter (thread GL 14) for venting the disposal can during filling. Available either in gray PP for a PP-coupling or in black PE for a stainless steel coupling.