Imprimer
Viresolve® Pro Device
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VPMD102NB1 VPMD103NB1
MILXVPMD102NB1EA 4420 EUR
MILXVPMD102NB1 MILXVPMD103NB1
Viresolve® Pro Device
Filtres Filtres à membrane
The Viresolve® pro solution provides a comprehensive, flexible template solution for viral clearance within mAb and recombinant protein bioproduction. This proven viral clearance solution delivers the highest levels of retention assurance and productivity across a broad range of feed stream characteristics.
  • Fully disposable single-use fluid path
  • Devices 100% tested with air/water diffusion and binary gas test
  • Shield/shield H 100% aerosol tested
Viresolve pro modus devices are typically used for filtration of pilot to mid-scale batch volumes of proteins. The Viresolve® Pro Device Modus 1,2 and 1,3 offers a filtration area of 0,07 and 0.22 m² and has a primary use in pilot/small-volume processing.