Imprimer
Stainless Steel Heavy Duty Tables with Multibar Undershelf
Fournisseur: TEKNOMEK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TLWB-MU/M12X60 TLWB-MU90X60 TLWB-MU60X60 TLWB-MU/M90X60 TLWB-MU/M60X60
TEKMTLWB-MU/M12X60EA 0 EUR
TEKMTLWB-MU/M12X60 TEKMTLWB-MU90X60 TEKMTLWB-MU60X60 TEKMTLWB-MU/M90X60 TEKMTLWB-MU/M60X60
Stainless Steel Heavy Duty Tables with Multibar Undershelf
Mobilier Tables
Stainless steel heavy duty table with multibar undershelf is manufactured with 304L-grade stainless steel. Crevice free design ensures suitability for hygienic industries.
  • 304L-grade stainless steel
  • Brushed dull polish finish
  • 4× stainless steel feet with non-marking floor levelling rubber base (static)
  • 4× stainless steel castors (2 swivel and 2 braked) (mobile)
  • Between shelves: 595 mm
  • Floor clearance to multibar: 175 mm
  • 250 kg payload (225 kg on mobile)
Stainless heavy duty table with multi bar undershelf is a very useful item for work surfaces or storage for industries that require sterile, hygienic and contamination resistant surfaces. It is more sturdy with a heavier payload for items that are of a heavier weight.
Finished in brushed dull polish, it comes in 304L-grade stainless steel or a customised 316L-grade stainless steel construction. Whichever you choose, the stainless steel heavy duty table will have a brushed dull polish finish. This stainless steel heavy duty table with multibar undershelf features a choice of adjustable feet with non-marking rubber base or stainless steel castors. The space underneath can also be utilised for extra storage.
Available in various widths, it features 175 mm ground clearance which enables easy cleaning and an overall height of 840 mm. Upstand version comes with inset legs (100 mm) in order to allow clearance if placed against a wall.