Imprimer
Metro® Super Erecta® Slide Systems
Fournisseur: Metro International
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SS2NC SS3NC
METOSS2NCEA 0 EUR
METOSS2NC METOSS3NC
Metro® Super Erecta® Slide Systems
Porte-lames
The Metro Super Erecta Slide System permits total adaptability of slides and tote boxes to any 530 or 610 mm wide Super Erecta Shelf ® open-wire unit.
  • Open-wire slide construction allows for ease of cleaning, on or off the unit
  • Super space-saving solution
  • Finish is nickel-chrome wire with clear protective coating
Slides are available in four sizes and can be positioned quickly and easily on new or existing units without the use of tools. Adaptable boxes are available in six sizes, with varying widths, lengths, and depths.
The Metro Super Erecta Slide System allows positioning of slides and tote boxes in vertical, horizontal, or freestyle arrangements, for maximum utilization of shelf space. 76, 152, and 203 mm box depths, in a variety of lengths and widths, hold a wide range of contents. The three different box depths, in different combinations, permit adjustment of shelf spacing from 305 to 483 mm.
Repositioning of slides avoids being 'locked in' to one arrangement, as with permanent vertical, or welded single slide systems. As requirements change, reposition the slides on the existing unit or move them to another Metro stationary or mobile unit.
Open wire, one-piece slides in four sizes are easily positioned or repositioned without tools. Slides install easily by snapping securely into place on overhead shelf.