Imprimer
SNAP i.d.® 2.0 Protein Detection Systems
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SNAP2MIDI SNAP2MINI SNAP2MM SNAP2MB2 SNAP2MB1 SNAP2MB3
MILFSNAP2MIDIEA 1900 EUR
MILFSNAP2MIDI MILFSNAP2MINI MILFSNAP2MM MILFSNAP2MB2 MILFSNAP2MB1 MILFSNAP2MB3
SNAP i.d.® 2.0 Protein Detection Systems
Analyseurs Analyseur de protéines
Developed to meet the western blotting customers, the SNAP i.d. 2.0 system produces blots of a very high quality. Unique vacuum-driven technology and a built-in flow distributor active.
  • Faster results
  • Faster testing of different antibodies
  • Higher throughput of Western blots each day
Informations de livraison:
This system includes the SNAP i.d. 2.0 base, two MultiBlot holding frames, two antibody collection trays, vacuum tubing, rolling Pad and blot roller, two wetting trays and a quick start user guide.