Imprimer
Storage Racks for Ultra-Low Temperature Freezers
Fournisseur: Thermo Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
920090 1950520 1950592 920091
HERA920090EA 258 EUR
HERA920090 HERA1950520 HERA1950592 HERA920091
Storage Racks for Ultra-Low Temperature Freezers
Portoirs Portoirs pour cryoboîtes
Cryobox storage racks makes organising your ultra-low temperature freezer samples easy.
  • Sliding drawer racks
  • Adjustable side access box racks
  • Stores 5 cm (2") and 7.6 cm (3") cryo boxes and microplates
Certifications:
cULus.