Imprimer
PYREX® Bottles, Media-lab, Wide Mouth, Clear, with Cap and Pouring Ring, DWK Life Sciences
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1523/06 1523/08 1523/10 1523/12 1523/14 1523/16 1523/18 1523/22
BIBB1523/06EA 159 EUR
BIBB1523/06 BIBB1523/08 BIBB1523/10 BIBB1523/12 BIBB1523/14 BIBB1523/16 BIBB1523/18 BIBB1523/22
PYREX® Bottles, Media-lab, Wide Mouth, Clear, with Cap and Pouring Ring, DWK Life Sciences
Flacons Flacons en verre
These bottles are a wide neck version of the standard GL 45 screw cap media-lab bottles. They are ideal for the storage of a wide selection of liquids, powders, and solids where a standard neck bottle would be too narrow.
  • GLS 80® screwthread provides increased access compared to GL 45 bottles, to aid filling and pouring
  • Printed with Trace Code for downloadable batch certificate
  • White enamel graduations and marking spot
  • Autoclavable
  • Screw caps and pouring rings can withstand temperatures up to 140 °C
Manufactured from PYREX® borosilicate glass with proven temperature and chemical resistance qualities.
Ideal for use with viscous liquids, pastes, powders, granules, and larger solids.