Imprimer
VWR® Rigid PCR Plates
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
732-4396EA 57.3 EUR
732-4396 732-4397 732-4398 732-4399 732-4400 732-4401 732-4402 732-4404 732-4406 732-4407 732-4408 732-4409 732-4410 732-4411 732-4412 732-4413 732-4414 732-4415 732-4416 732-4417 732-4418 732-4419 732-4421 732-4423 732-4424
VWR® Rigid PCR Plates
Microplaques Plaques PCR
VWR® Rigid PCR Plates feature a rugged polycarbonate frame, providing a solid and flat surface that guarantees seamless interaction with automated liquid handling platforms. With a firm grip assured, robots can confidently place VWR® Rigid PCR Plates with utmost precision, eliminating any concerns of misalignment.
  • Polycarbonate frame is rigid and flat to facilitate a firm grip by robotic arms and ensure reliable placement
  • Clear polypropylene wells provide optimal thermal transfer and higher reaction efficiency
  • White well options are available to enhance the fluorescence signal during qPCR (search VWR Rigid qPCR Plates)
  • Wells are slightly raised to ensure evaporation-proof sealing with film, foil or strip caps
  • ANSI/SBS footprint and well spacing
  • Vibrant colours offer a visual distinction between different samples or experiments
VWR® Rigid PCR Plates feature no-flex polycarbonate frames and polypropylene wells designed to facilitate optimal thermal transfer and higher reaction efficiency. This ensures more accurate and reliable results, providing researchers with the confidence they need in their experiments. Additionally, our white well options are carefully designed to enhance fluorescence signals during qPCR, enabling clearer and more precise measurements.
Sealing PCR plates is a critical aspect of laboratory workflows, and VWR® excels in this area. The slightly raised wells of VWR® Rigid PCR Plates ensure an evaporation-proof seal when used with film, foil or strip caps. Say goodbye to concerns about sample integrity or loss due to evaporation- VWR® Rigid PCR Plates keep your valuable samples protected throughout the entire process.
With an ANSI/SBS footprint and well spacing, VWR® Rigid PCR Plates integrate seamlessly into existing laboratory setups, offering compatibility and ease of use. Plus, our vibrant colour options allow quick and efficient identification and organisation by providing visual distinctions between different samples or experiments.
Certifications:
Certified free of RNase, DNase, DNA and PCR Inhibitors.
Cdt:
10 plates per pack, 10 packs per case (total 100 plates).