Imprimer
Scepter™ 3.0 Automated Cell Counter Kits
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PHCC360KIT PHCC360050 PHCC340050 PHCC360250 PHCC340250 PHCC340500 PHCC360500 PHCC340KIT
MILFPHCC360KITEA 4390 EUR
MILFPHCC360KIT MILFPHCC360050 MILFPHCC340050 MILFPHCC360250 MILFPHCC340250 MILFPHCC340500 MILFPHCC360500 MILFPHCC340KIT
Scepter™ 3.0 Automated Cell Counter Kits
Compteurs de cellules et cytomètres de flux Compteurs de cellules automatisés
The Scepter™ 3.0 handhled automated cell counter provides the unparalleled precision of Coulter impedance counting in a convenient, handheld format. Newest version of the Scepter™ cell counter has been updated with features that make it more convenient, easy-to-use, and accurate.
  • Coulter impedance method which avoids common artifacts arising from image-based cell counting methods
  • Does not rely on user technique or manual calculation for accuracy
  • Counts thousands of cells per measurement for maximum precision
  • Unique ergonomic design, convenient for measurements and storage at the tissue culture hood
  • Returns precision counts in <30 seconds
  • Requires no sample prep, dedicated reagents, or hazardous dyes
  • Requires no cleaning protocol to maintain operation
Can be used to monitor cell health between measurements, passages and batches using size and morphology.
The Scepter™ sensors use precision microfluidics to measure the electrical impedance of individual cells. This technology gives on-demand outputs of cell volume, cell diameter and cell concentration all within the cell culture hood.
40 µm sensor validated performance range
Operating range: 50000 to 1500000 cells/ml
Sensor scale diameter: 4 to 20 µm
Working diameter range: 5 to 15 µm
60 µm sensor
Operating range: 10000 to 500000 cells/ml
Sensor scale diameter: 6 to 36 µm
Working diameter range: 8 to 25 µm
Informations de livraison:
Each kit includes: Handheld automated cell counter, cell counter sensors (50 pack), charger station and mounting kit, test bead vial, quick start guide and safety information.