Imprimer
WHEATON® Weighing Boats, Diamond Shape
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DS30B/AS DS5B/AS
LSUKDS30B/ASEA 280 EUR
LSUKDS30B/AS LSUKDS5B/AS
WHEATON® Weighing Boats, Diamond Shape
Nacelles Nacelles de pesée
WHEATON® weighing boats are anti-static, available in a diamond shape to suit most user applications.
  • Manufactured from high-impact polystyrene
  • Ideal for handling both liquids and solids
Anti-static design prevents retention of sample by static electricity. Rounded corners provide for easy removal of powdered and granular samples.