Imprimer
Thermo Scientific™ TSG Undercounter Refrigerators
Fournisseur: Thermo Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TSG505GA TSG505SA
HERATSG505GAEA 4050 EUR
HERATSG505GA HERATSG505SA
Thermo Scientific™ TSG Undercounter Refrigerators
Réfrigérateurs
TSG Series, powered by solid state refrigeration technology, has been designed to meet the rigorous and demanding needs of healthcare, research and industrial applications where solid performance is required.
  • Solid state refrigeration technology
  • Whisper quiet operation leading to a more comfortable work environment supporting clinical and healthcare needs
  • Increased capacities due to fewer refrigeration components means you can store more in less space
  • Energy Star rated performance meaning lower operating costs
  • Green refrigerants supporting global
En savoir plus