Imprimer
Liquid Ends for Liquid Handling Station
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
709410 709413 709420 709423 709416 709426
BRND709410EA 850 EUR
BRND709410 BRND709413 BRND709420 BRND709423 BRND709416 BRND709426
Liquid Ends for Liquid Handling Station
Automated Liquid Handling Systems
The basis for the pipette modules are components from the familiar Transferpette® S.
  • Autoclavable at 121 °C for 20 minutes
Three single-channel Liquid Ends (SC) and three 8-channel Liquid Ends (MC) are available for contact-free liquid delivery. The volume testing of the pipette modules is carried out according to ISO 8655 part 6.