Imprimer
Cooling Blocks, Passive Cooler
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
709510 709511 709512 709513 709514
BRND709510EA 1020 EUR
BRND709510 BRND709511 BRND709512 BRND709513 BRND709514
Cooling Blocks, Passive Cooler
Chauffe-blocs Blocs pour bains
Keep your samples and media cool for up to three hours with these Cooling Blocks from BRAND.
  • ANSI/SLAS format for use in pipetting robotic systems such as the Liquid Handling Station
  • Cools microcentrifuge tubes and PCR consumables
All well positions are cooled homogenously. The Cooling Blocks work as a passive heat sink which makes them quick and easy to use without an additional power supply.