Imprimer
TriLink® 2'-Deoxyadenosine-5'-Triphosphate
Marques : TriLink BioTechnologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N-2506-100 N-2506-25
SOLUN-2506-100EA 206 EUR
SOLUN-2506-100 SOLUN-2506-25
TriLink® 2'-Deoxyadenosine-5'-Triphosphate
Réactifs pour acides nucléiques Nucléotides
This nucleotide consists of an unmodified base, a deoxyribose, and a 5′ triphosphate group for synthesis of DNA molecules.
  • Unmodified dexoyribonucleotide for DNA synthesis
  • ≥97% purity by AX-HPLC analysis
  • Provided as 100 mM in H₂O, pH 7,5
  • Grade: RUO
Deoxyadenosine-5′-triphosphate (dATP), deoxycytidine-5′-triphosphate (dCTP), deoxyguanosine-5′-triphosphate (dGTP), and deoxythymidine-5′-triphosphate (dTTP) are unmodified deoxyribonucleoside triphosphates, serving as essential building blocks of DNA molecules. Each of them consists of an unmodified base, a dexoyribose, and a 5′ triphosphate group.
Attention: Item will be delivered at or below −20 °C and should be stored at or below −20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10972a1bf7c5030dd990a85ff2d193c393c5c809&vid=10972a1bf7c5030dd990a85ff2d193c393c5c809&prid=web&clid=SAPDAM