Imprimer
TriLink® Biotin-16-Aminoallyluridine-5'-Triphosphate
Marques : TriLink BioTechnologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N-5005-0250 N-5005-1 N-5005-2
SOLUN-5005-0250EA 196 EUR
SOLUN-5005-0250 SOLUN-5005-1 SOLUN-5005-2
TriLink® Biotin-16-Aminoallyluridine-5'-Triphosphate
Réactifs pour acides nucléiques Nucléotides
Biotin-16-Aminoallyluridine-5'-Triphosphate (Biotin-16-UTP, Biotin-16-AA-UTP) can be incorporated via a number of RNA polymerases, including SP6-, T3- and T7-RNA polymerase to form labeled RNA.
  • Grade: RUO
According to Fenn BJ, et al., T7 RNA polymerase does not significantly discriminate between the natural and the biotinylated nucleotide. Therefore, the number of biotinylated positions per transcript can be controlled by varying the ratio of the biotinylated NTP to the standard NTP. Biotinylated RNA probes are useful as a nonradioactive label in a number of applications including chromosome mapping, in situ hybridisation and microarray hybridisation. Biotin-16-U-modified RNA can be detected with a variety of streptavidin-based approaches.
Attention: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=806bd3c9b6fbee08ece6ef6c718cfc129024df25&vid=806bd3c9b6fbee08ece6ef6c718cfc129024df25&prid=web&clid=SAPDAM