Imprimer
TriLink® CleanAmp® 7-deaza-dGTP Mix
Marques : TriLink BioTechnologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N-9504-10 N-9504-2
SOLUN-9504-10EA 615 EUR
SOLUN-9504-10 SOLUN-9504-2
TriLink® CleanAmp® 7-deaza-dGTP Mix
Réactifs pour acides nucléiques Nucléotides
CleanAmp® dNTPs are chemically-modified nucleotides designed to improve specificity, sensitivity, and robustness of your PCR.
  • Improves PCR specificity, sensitivity, and robustness
  • Provides an alternative approach to hot-start PCR without a need for a specially designed DNA polymerase
  • Grade: RUO
The CleanAmp® 7-deaza-dGTP: - Contains CleanAmp® 7-deaza deoxyguanosine 5’ triphosphate in a 2 µmol or 10 µmol format - Is ideal for GC-rich PCR (~60% GC) due to promoting the template’s strand separation by lowering its melting temperature - Enables hot-start PCR for specificity and sensitivity, with a thermolabile protecting group at the 3’ hydroxyl of the nucleotides (CleanAmp® dNTP benefit) - Has been QC analysed by AX-HPLC, RP-HPLC, 31P NMR, mass spec, and a standardised PCR assay
Attention: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5084b3b0e0412f89888e99c31de45dc5de5d06af&vid=5084b3b0e0412f89888e99c31de45dc5de5d06af&prid=web&clid=SAPDAM
New Product
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Application
Learn more