Imprimer
TriLink® Uridine-5'-Triphosphate
Marques : TriLink BioTechnologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N-1504-10 N-1504-100 N-1504-25
SOLUN-1504-10EA 25.3 EUR
SOLUN-1504-10 SOLUN-1504-100 SOLUN-1504-25
TriLink® Uridine-5'-Triphosphate
Réactifs pour acides nucléiques Nucléotides
This nucleotide consists of an unmodified base, a ribose, and a 5′ triphosphate group for synthesis of RNA molecules.
  • Unmodified ribonucleotide for RNA synthesis
  • >99% purity by AX-HPLC analysis
  • Provided as 100 mM in H₂O, pH 7,5
  • Grade: RUO
Attention: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5677931d76e5a1c48f329559f5931df1b29fb933&vid=5677931d76e5a1c48f329559f5931df1b29fb933&prid=web&clid=SAPDAM