Imprimer
TriLink® 5-Methylcytidine-5'-Triphosphate
Marques : TriLink BioTechnologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N-1014-10 N-1014-1 N-1014-5
SOLUN-1014-10EA 496 EUR
SOLUN-1014-10 SOLUN-1014-1 SOLUN-1014-5
TriLink® 5-Methylcytidine-5'-Triphosphate
Réactifs pour acides nucléiques Nucléotides
This nucleotide consists of a modified base, a ribose, and a 5′ triphosphate group.
  • Can be incorporated in in vitro transcription for modification of synthesised RNA for desired characteristics
  • ≥95% purity by AX-HPLC analysis, provided as 100 mM in H₂O
  • Grade: RUO
Methylation of the 5 position of cytidine is a common, post-transcriptional modification in a number of RNA species, such as mRNA, miRNA and tRNA. 5-Methylcytidine-5'-Triphosphate (5-Methyl-CTP) is a modified nucleoside triphosphate employed to impart desirable characteristics such as increased nuclease stability, increased translation or reduced interaction of innate immune receptors with in vitro transcribed RNA.
Attention: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.