Imprimer
TriLink® N6-Methyladenosine-5'-Triphosphate
Marques : TriLink BioTechnologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N-1013-1 N-1013-10 N-1013-5
SOLUN-1013-1EA 94.3 EUR
SOLUN-1013-1 SOLUN-1013-10 SOLUN-1013-5
TriLink® N6-Methyladenosine-5'-Triphosphate
Réactifs pour acides nucléiques Nucléotides
N6-methyl adenosine (N6-methyl ATP) is a base modified analog of adenosine and is found as a minor nucleoside in natural RNAs (Meyer et al.). N6-methyl ATP can substitute for ATP in some biological systems, and is a potent agonist for P2Y-purinoceptors in the guinea pig, taenia coli (Burnstock et al.).
  • Grade: RUO
Attention: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.