Imprimer
TriLink® CleanCap® Reagent AG
Marques : TriLink BioTechnologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N-7113-10 N-7113-1 N-7113-5 N-7113-100
SOLUN-7113-10EA 1070 EUR
SOLUN-7113-10 SOLUN-7113-1 SOLUN-7113-5 SOLUN-7113-100
TriLink® CleanCap® Reagent AG
Réactifs pour acides nucléiques RNA Synthesis and Transcription
CleanCap® Reagent AG is TriLink’s patented co-transcriptional capping reagent for in vitro transcription of 5’ capped mRNA resulting in a Cap 1 structure. CleanCap has shown to provide up to 98% capping efficiency.
  • Grade: RUO
PLEASE NOTE: CleanCap Reagent AG requires an AG initiator. To learn more, please review the Product Insert.
Attention: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=658e4421b672768efb61f79ae1faca3a36a4c1d3&vid=658e4421b672768efb61f79ae1faca3a36a4c1d3&prid=web&clid=SAPDAM