Imprimer
Green Cellulose Sterilisation Wraps, Keystone Cleanroom Products
Fournisseur: KEYSTONE CLEANROOM PRODUCTS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
GHDSHEET01.5 GHDSHEET0202 GHDSHEET0303 GHDSHEET1818 GHDSHEET2020 GHDSHEET0404 GHDSHEET0606 GHDSHEET0707 GHDSHEET0808 GHDSHEET0909 GHDSHEET1212 GHDSHEET1515 GHDSHEET5472 GHDSHEET6060 GHDSHEET6080 GHDSHEET2424 GHDSHEET3030 GHDSHEET3636 GHDSHEET4040 GHDSHEET4545 GHDSHEET4548 GHDSHEET4848 GHDSHEET5454
KACCGHDSHEET01.5EA 215 EUR
KACCGHDSHEET01.5 KACCGHDSHEET0202 KACCGHDSHEET0303 KACCGHDSHEET1818 KACCGHDSHEET2020 KACCGHDSHEET0404 KACCGHDSHEET0606 KACCGHDSHEET0707 KACCGHDSHEET0808 KACCGHDSHEET0909 KACCGHDSHEET1212 KACCGHDSHEET1515 KACCGHDSHEET5472 KACCGHDSHEET6060 KACCGHDSHEET6080 KACCGHDSHEET2424 KACCGHDSHEET3030 KACCGHDSHEET3636 KACCGHDSHEET4040 KACCGHDSHEET4545 KACCGHDSHEET4548 KACCGHDSHEET4848 KACCGHDSHEET5454
Green Cellulose Sterilisation Wraps, Keystone Cleanroom Products
Lingettes de stérilisation
When secured with autoclave tape or elasticised bands, these green GHD wrappers provide an effective microbial barrier from external sources of contamination.
  • Designed for steam sterilisation; but also compatible with ETO or gamma irradiation
  • Treated to impart water and alcohol repellency
  • Wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibers bonded with a resin binder
  • Sufficient microbial barrier
  • Superior bacterial barrier properties compared to linen
  • Sterilization tapes adhere firmly and are easy to remove
  • Ideal choice for light trays, small basins and towels
  • Resist tearing and punctures
  • Do not contain any natural latex rubber
  • Processed and packaged in ISO certified cleanrooms
Informations de livraison:
Certificates of Conformance sent with each shipment.
Cdt:
Packaged in sealed, labeled poly bags with lot number and date of manufacture.